Product Image
Thermo Scientific™

CEDIA™ Benzodiazepine Assays

Utilize the globally valued, highly accurate, and precise Thermo Scientific™ CEDIA™ Benzodiazepine Assay for very stable shelf life and lot-to-lot dependability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
17755611 x 495 mL, LC
1000853 x 17 mL, TSC
100164093 x 17 mL, Indiko
1000941 x 65 mL, MCC
Catalog number 1775561
Price (INR)
-
Quantity:
1 x 495 mL, LC

Utilize the globally valued, highly accurate, and precise Thermo Scientific™ CEDIA™ Benzodiazepine Assay for very stable shelf life and lot-to-lot dependability. Applications are available for an array of clinical chemistry analyzers. The β-Glucuronidase Enzyme, when used with this assay, offers enhanced sensitivity of parent drug by hydrolyzing the glucuronide metabolites. The Thermo Scientific CEDIA panel of drugs of abuse assays is ever expanding to address emerging drug trends and testing needs.

  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Benzodiazepine Assays
Quantity1 x 495 mL, LC
Sample TypeUrine
TypeReagent
Detectable AnalytesNitrazepam
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C