6-Methoxyquinoline, 98%
6-Methoxyquinoline, 98%
6-Methoxyquinoline, 98%
Acros Organics

6-Methoxyquinoline, 98%

CAS: 5263-87-6 | C10H9NO | 159.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
179340250
also known as 17934
25 gGlass bottle
Catalog number 179340250
also known as 17934
HSN 29334900
Price (INR)
10,889.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
10,889.00
Each
Chemical Identifiers
CAS5263-87-6
IUPAC Name6-methoxyquinoline
Molecular FormulaC10H9NO
InChI KeyHFDLDPJYCIEXJP-UHFFFAOYSA-N
SMILESCOC1=CC=C2N=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or low melting solid
Refractive index1.6220 to 1.6260 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear yellow to pink to purple to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only