Product Image
Thermo Scientific™
Accucore™ Narrow Selectivity Kits
Verify optimum selectivity of Thermo Scientific™ Accucore™ HPLC columns over a narrow range.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
17X26-102130-3VA100 mm
17X26-152130-3VA150 mm
Catalog number 17X26-102130-3VA
Price (INR)
-
Length (Metric):
100 mm
Verify optimum selectivity of Thermo Scientific™ Accucore™ HPLC columns over a narrow range with Accucore Narrow Selectivity Kits.
Based on Core Enhanced Technology™, Accucore columns provide fast, high-resolution separations without the elevated backpressures required by sub-2μm particles.
  • Fast separations with excellent resolution and sensitivity
  • Lower backpressures so that UHPLC is not required
  • Kit contains 1 x Accurcore C18 HPLC Column, 1 x Accucore RP-MS Column, 1 x Accucore aQ Column
Specifications
Column FormatHPLC Column Kit
Column TypeReversed Phase
EndcappedYes
For Use WithHPLC
Particle ShapeSpherical Solid Core
Stationary PhaseaQ C18, C18, RP-MS
Surface Area130 m²/g
Diameter (Metric)2.1 mm
Length (Metric)100 mm
Particle Size2.6 μm
PhaseReversed Phase
Pore Size80 Å
Unit SizeEach