Product Image
Invitrogen™

eBioscience™ Avidin HRP

Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoproteinRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
18-4200-93100 μL
18-4200-895 x 100 μL
Catalog number 18-4200-93
Price (INR)
15,540.00
Online offer
Ends: 31-Mar-2026
25,899.00
Save 10,359.00 (40%)
Each
Quantity:
100 μL
Price (INR)
15,540.00
Online offer
Ends: 31-Mar-2026
25,899.00
Save 10,359.00 (40%)
Each

Avidin horseradish peroxidase is commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoprotein from chicken egg whites that binds biotin with high affinity. Biotin is a tag that is frequently used to label antibodies and other probes for immunodetection methods.

This product is recommended for use in immunohistochemistry applications.

Conjugate: HRP

Reported Application: Immunohistochemical Staining

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormulationAqueous buffer, contains carrier protein/stabilizer, no sodium azide
Label or DyeHRP (Horseradish Peroxidase)
Quantity100 μL
Target MoleculeBiotin
For Use With (Application)Immunohistochemical Staining
FormLiquid
Product LineeBioscience
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C