18mm Headspace Vials & Closures
Actual product may vary

18mm Headspace Vials & Closures

Vials are available in 10mL and 20mL usable volumes with screw thread closure styles that require no tools for closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorClosure TypeMaterialProduct Type
18-MSC-ST101Blue, SilverMagnetic Screw Cap (Open-Top)PTFE, Silicone, SteelMagnetic Screw Cap
20-HSVClearScrew CapGlass18 mm Screw Top Headspace Vial
18-MSC
also known as CHROMACOL 18-MSC
SilverMagnetic Screw Cap (Open-Top)SteelMagnetic Screw Cap
18-MSC-ST3HT
also known as THERMO GERMANY 18-MSC-ST3HT
Silver, RedScrew Cap (Magnetic) with SeptumPTFE, Silicone, Steel
18-ST101BlueSeptum OnlyPTFE, SiliconeSeptum
18-MSC-ST201Silver, BlueScrew Cap (Magnetic) with SeptumSilicone, PTFE, Steel
Catalog number 18-MSC-ST101
HSN 83099090
Price (INR)
23,058.00
Each
Color:
Blue, Silver
Closure Type:
Magnetic Screw Cap (Open-Top)
Material:
PTFE, Silicone, Steel
Product Type:
Magnetic Screw Cap
Price (INR)
23,058.00
Each
Unlike crimp cap vials, Thermo Scientific™ National 18mm Screw Top Headspace Vials require no tools for closure. Manufactured with superior quality (Type 1, Class A) glass, they are available with a round bottom in sizes with usable volumes of 10mL or 20mL. The 18mm vials feature Magnetic Screw Caps which enable magnetic pick up in autosamplers with that capability. Multiple turn threading maintains a tight seal through extreme heating cycles. Septa are Blue Silicone/ PTFE.

Choose septa materials to meet your needs.

  • Gray Butyl Stoppers are an economical septum for lower temperature (125°C) or low-pressure applications.
  • Gray PTFE/Red Rubber Septa are an economical choice where a PTFE barrier is desired.
  • PTFE/White Silicone PurePack Septa for general volatiles analysis.
  • Gray PTFE/Molded Black Butyl Septa offer a more positive seal against loss of fixed gases.
  • PTFE/Blue High Purity Silicone Septum reduces background from extractables or outgassing of volatile contaminants.
  • The Black Rubber septum is an economical choice for applications where reduced levels of vapor penetration are desired.

Unassembled convenience kits
National and Chromacol Headspace Unassembled Convenience Kits save time during sample preparation and include matched quantities of vials and silver aluminum seals with prefitted septa for added convenience.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorSilver
Description18mm Magnetic Screw Cap, 8mm hole
Diameter (Metric) Opening18 mm
Diameter (Metric) Inner18 mm
ColorBlue, Silver
MaterialPTFE, Silicone, Steel
Septum18 mm Blue Silicone/PTFE, not prefitted
Closure MaterialSteel
Closure TypeMagnetic Screw Cap (Open-Top)
Quantity125
Thickness (Metric)1.0 mm
Closure Size18 mm
Green FeaturesACT Ecolabel (Silver)
Product TypeMagnetic Screw Cap
Unit SizeEach