2-Bromobenzyl alcohol, 98%
2-Bromobenzyl alcohol, 98%
2-Bromobenzyl alcohol, 98%
Acros Organics

2-Bromobenzyl alcohol, 98%

CAS: 18982-54-2 | C7H7BrO | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 180100100
also known as 18010
HSN 29062990
Price (INR)
7,480.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,480.00
Each
Chemical Identifiers
CAS18982-54-2
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point78°C to 82°C
GC>=97.5 %
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or needles
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only