LN<sub>2</sub> Measuring Stick
Thermo Scientific™
LN2 Measuring Stick
Use this measuring stick with cryopreservation equipment.
Have Questions?
Catalog NumberFor Use With (Equipment)
180143Locator, Locator Plus, CryoPlus
Catalog number 180143
Price (INR)
-
For Use With (Equipment):
Locator, Locator Plus, CryoPlus
Use Thermo Scientific™ LN2 Measuring Stick with our LN2 cryopreservation equipment.
Specifications
DescriptionLN2 Measuring Stick
For Use With (Equipment)Locator, Locator Plus, CryoPlus
TypeLN2 Measuring Device
Unit SizeEach
Frequently asked questions (FAQs)
How long is the LN2 Measuring Stick?
The LN2 measuring stick has a total length of ~114 cm (~45 inches) and the measuring portion of the stick is scaled to ~100 cm (40+ inch).