Isopropyl acetate, 99+%, pure
Isopropyl acetate, 99+%, pure
Isopropyl acetate, 99+%, pure
Acros Organics

Isopropyl acetate, 99+%, pure

CAS: 108-21-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Catalog number 180800025
also known as 18080
HSN 29153910
Price (INR)
16,570.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (INR)
16,570.00
Each
Chemical Identifiers
CAS108-21-4
IUPAC Namepropan-2-yl acetate
Molecular FormulaC5H10O2
InChI KeyJMMWKPVZQRWMSS-UHFFFAOYSA-N
SMILESCC(C)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only