SuperScript™ IV RT Reaction Buffer
Invitrogen™

SuperScript™ IV RT Reaction Buffer

SuperScript IV RT Reaction Buffer is included with, and designed for use with, SuperScript IV Reverse Transcriptase.
Have Questions?
Catalog NumberQuantity
18090050B5 x 1 mL
Catalog number 18090050B
Price (INR)
17,558.00
Online offer
Ends: 31-Dec-2026
25,082.00
Save 7,524.00 (30%)
Each
Quantity:
5 x 1 mL
Price (INR)
17,558.00
Online offer
Ends: 31-Dec-2026
25,082.00
Save 7,524.00 (30%)
Each

SuperScript IV RT Reaction Buffer is included with, and designed for use with, SuperScript IV Reverse Transcriptase. This buffer is optimized to give robust and reliable cDNA synthesis. In cases where additional quantity is required, SuperScript IV RT Reaction Buffer is made available here in a stand-alone format.

For use with

SuperScript IV Reverse Transcriptase (Cat. No 18090010)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Includes

• SuperScript IV RT Reaction Buffer (5 x 1 mL)

Store at –20°C.

Concentration5X
For Use With (Application)Reverse Transcription
Quantity5 x 1 mL
Unit SizeEach

Frequently asked questions (FAQs)

Can I use the SuperScript IV RT Reaction Buffer with SuperScript II RT or SuperScript III RT enzymes?

No, we do not recommend using SuperScript IV RT Reaction Buffer with the SuperScript II RT or SuperScript III RT enzymes. For optimal performance, we recommended using the matching reaction buffer for each enzyme.