Product Image
Invitrogen™
BioPrime™ Total Genomic Labeling System
The BioPrime™ Total Genomic Labeling System is a complete genomic DNA labeling system designed for use in array comparative genomicRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
18097011
also known as 18097-011
30 Reactions
1809701010 Reactions
Catalog number 18097011
also known as 18097-011
Price (INR)
135,805.00
Each
Quantity:
30 Reactions
Price (INR)
135,805.00
Each
The BioPrime™ Total Genomic Labeling System is a complete genomic DNA labeling system designed for use in array comparative genomic hybridization applications (aCGH) that allows for better call rates with precious samples.
Note: The BioPrime™ Total Genomic Labeling System includes PurelinkTM Genomic DNA purification columns and purification buffers. The BioPrime™ Total Genomic Labeling Module does not.
The BioPrime™ Total Genomic Labeling System offers users the following advantages:
- Higher signal to noise with Alexa Fluor™ 3 and 5 dyes
- Less channel bias due to novel reaction formulation
- Widest range of input material (50 ng - 3 μg)
- Simplified workflow with master mix formulation
This complete, all-inclusive kit is optimized to work across the widest range of sample input material with no need for pre-amplification. Highly concentrated exo-Klenow fragment of DNA polymerase I in a component-limited reaction allows for consistent, robust DNA yields of ∼ 8 μg with as little as 50 ng of genomic DNA input. An optimized dye-labeled nucleotide mix with new dNTP linkers, novel application-specific Alexa Fluor™ 3 and 5 dyes, and improved buffer chemistry reduce labeling variation and increase signal to noise on arrays. Excitation and emission spectra of Alexa Fluor™ 3 and 5 dyes are suited for conventional two color scanners with no need to change settings. New master mix formulation streamlines reaction set-ups, increasing consistency. Complete kits are batch tested to assure quality in performance from lot to lot.
Contents and Storage:
The BioPrime™ Total Genomic Labeling System contains Alexa Fluor™ 3 and Alexa Fluor™ 5 2x reaction mix, Exo-Klenow fragment (40 U/ μl), 5 mM EDTA, TE Buffer, and control DNA (Salmon Sperm). Store at -80°C for long term storage. The 2X Reaction Mixes may be stored at +4°C for up to 4 weeks and should be protected from light. The BioPrime™ Total Genomic Labeling System also contains PurelinkTM Genomic DNA purification columns and purification buffers (the BioPrime™ Total Genomic Labeling Module does not). Store at room temperature. All components are guaranteed for 6 months when properly stored.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Includes Label or DyeYes
Labeling MethodDirect Labeling
Product LineBioPrime
Product TypeTotal Genomic Labeling System
Quantity30 Reactions
Detection MethodFluorescence
Final Product TypeProbes (Labeled DNA)
Labeling TargetGenomic DNA
Label or DyeAlexa Fluor 3, Alexa Fluor 5
Unit SizeEach
Frequently asked questions (FAQs)
Why do you need more labeling product for a cDNA array than for a BAC array?
The reason for using more genomic DNA for the cDNA array is because the BAC arrays have much longer DNA fragments on the array than cDNA, therefore higher homology to the labeled DNA than the cDNA array. The fluorescence signals from the cDNA will be weaker than the BAC array, therefore, you have to start with higher amounts of starting materials.
Why is the selection of a BioPrime Genomic Labeling System so important?
The overall performance of your microarray experiment is driven by numerous factors including the microarray, amplification method, labeling method, hybridization solutions/conditions, blocking and wash methods, as well as performance of the associated hardware systems. In order to achieve the highest confidence in your array results best in class components must be selected for each step. Invitrogen’s BioPrime Total and Cot-1 DNA products are the proven labeling and blocking reagents to increase accuracy, sensitivity, and simplicity.
When using BioPrime Genomic Labeling Systems, why is random primed linear amplification (RPA) so important for these applications?
All of the BioPrime Genomic Labeling systems are based on linear, random primed amplification methods-the best method to maintain accurate representation of copy number. Other methods of amplification may provide inaccurate amplification of starting material which potentially invalidate array results.
What are the differences between Alexa Fluor 3 and 5 used in the BioPrime Genomic Labeling Systems?
Both lines of Alexa Fluor dyes can be used on traditional 2 color scanners with excitation and emission wavelengths optimized for these scanners.  The Alexa Fluor 3 and Alexa Fluor 5 dye conjugated nucleotides have been optimized for DNA Microarray applications and are incorporated with the enzymes used in BioPrime Total Genomic Labeling systems to provide excellent performance when dried down on arrays. Excitation/Emission for Alexa Fluor 3 is 555/565 nm. For Alexa Fluor 5 it is 650/670 nm.
Which BioPrime Genomic Labeling System is right for me?
The BioPrime Total array CGH reagents offer various configurations to allow for optimal performance on arrays based on premier enzymes, primers, dyes and purification systems. For 2 color array CGH and ChIP on Chip experiments Invitrogen recommends the BioPrime Total Genomic Labeling system. The BioPrime Total Genomic Labeling system allows for the lowest starting input material, includes application optimized dye labeled nucleotides with Alexa Fluor 3 and Alexa Fluor 5 dyes, and is the simplest, easiest to use kit in the product family.  For researchers favoring other dyes, BioPrime aCGH Genomic Labeling systems are available without dyes and can be used with your dye of preference to offer superior yield and total dye incorporation due to higher performance enzymes and reagents from Invitrogen. Both BioPrime Total and BioPrime array CGH kits have been successfully used with both oligo and BAC arrays from vendors such as:  Agilent, Perkin Elmer, Signature Genomics, Combimatrix Diagnostics, and BlueGnome. For microarray systems requiring biotin labeling the BioPrime DNA Labeling system includes biotin labeled nucleotides in the kit and has been successfully used in both one and two color microarray experiments. View the Genomic array CGH selection guide.
Citations & References (1)
Citations & References
Abstract
Random DNA fragmentation allows detection of single-copy, single-exon alterations of copy number by oligonucleotide array CGH in clinical FFPE samples.
Authors:Hostetter G, Kim SY, Savage S, Gooden GC, Barrett M, Zhang J, Alla L, Watanabe A, Einspahr J, Prasad A, Nickoloff BJ, Carpten J, Trent J, Alberts D, Bittner M,
Journal:Nucleic Acids Res
PubMed ID:19875416
'Genomic technologies, such as array comparative genomic hybridization (aCGH), increasingly offer definitive gene dosage profiles in clinical samples. Historically, copy number profiling was limited to large fresh-frozen tumors where intact DNA could be readily extracted. Genomic analyses of pre-neoplastic tumors and diagnostic biopsies are often limited to DNA processed by ... More