1,10-Decanediol, 99%
1,10-Decanediol, 99%
1,10-Decanediol, 99%
Acros Organics

1,10-Decanediol, 99%

CAS: 112-47-0 | C10H22O2 | 174.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 181041000
HSN 29053990
Price (INR)
7,054.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,054.00
Each
Chemical Identifiers
CAS112-47-0
IUPAC Namedecane-1,10-diol
Molecular FormulaC10H22O2
InChI KeyFOTKYAAJKYLFFN-UHFFFAOYSA-N
SMILESOCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or flakes or pellets
Infrared spectrumConforms
Melting point72°C to 74°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only