N,N-Diethylaniline, 99%
N,N-Diethylaniline, 99%
N,N-Diethylaniline, 99%
Acros Organics

N,N-Diethylaniline, 99%

CAS: 91-66-7 | C10H15N | 149.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number 181060025
also known as 18106
HSN 29214290
Price (INR)
12,404.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,404.00
Each
Chemical Identifiers
CAS91-66-7
IUPAC NameN,N-diethylaniline
Molecular FormulaC10H15N
InChI KeyGGSUCNLOZRCGPQ-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5410 to 1.5430 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only