3,5-Dimethylpiperidine, 96%, mixture of cis and trans
3,5-Dimethylpiperidine, 96%, mixture of cis and trans
3,5-Dimethylpiperidine, 96%, mixture of cis and trans
Acros Organics

3,5-Dimethylpiperidine, 96%, mixture of cis and trans

CAS: 35794-11-7 | C7H16N | 114.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 181231000
HSN 29333919
Price (INR)
39,295.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
39,295.00
Each
Chemical Identifiers
CAS35794-11-7
IUPAC Name(3R,5S)-3,5-dimethylpiperidin-1-ium
Molecular FormulaC7H16N
InChI KeyIDWRJRPUIXRFRX-KNVOCYPGSA-O
SMILESC[C@H]1C[NH2+]C[C@@H](C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=95 % (sum of cis and trans)
Refractive index1.4425 to 1.4475 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only