Gluconic acid, sodium salt, 98%
Gluconic acid, sodium salt, 98%
Gluconic acid, sodium salt, 98%
Acros Organics

Gluconic acid, sodium salt, 98%

CAS: 527-07-1 | C6H11NaO7 | 218.137 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 181390050
HSN 29181690
Price (INR)
2,864.00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
2,864.00
Each
Chemical Identifiers
CAS527-07-1
IUPAC Namesodium 2,3,4,5,6-pentahydroxyhexanoate
Molecular FormulaC6H11NaO7
InChI KeyUPMFZISCCZSDND-UHFFFAOYNA-M
SMILES[Na+].OCC(O)C(O)C(O)C(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Titration with HClO4>=97.5 %
Chloride (Cl)=<100 ppm
Reducing substances=<0.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only