Search
Search
| Catalog Number | Quantity | Type |
|---|---|---|
| 1815334 | 1 x 15 mL | Primary Calibrator |
| 1730380 | 1 x 5 mL | Intermediate Calibrator |
| 1730398 | 1 x 5 mL | High Calibrator |
| 1730401 | 1 x 5 mL | Clinical Calibrator |
| 1730428 | 1 x 5 mL | Secondary Calibrator |
| 1730517 | 1 x 15 mL | Secondary Calibrator |
| 1732218 | 1 x 15 mL | Intermediate Calibrator |
| 1732226 | 1 x 15 mL | High Calibrator |
| 1815326 | 1 x 5 mL | Primary Calibrator |
Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Multidrug Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.