Product Image
Invitrogen™

DNA Polymerase I/DNase I

DNA Polymerase I/DNase I is an optimized mixture of both enzymes for efficient nick translation of DNA.Application: Labeling DNA withRead more
Have Questions?
Catalog NumberQuantity
18162016250 U
Catalog number 18162016
Price (INR)
18,291.00
Online offer
Ends: 31-Dec-2026
30,485.00
Save 12,194.00 (40%)
Each
Quantity:
250 U
Price (INR)
18,291.00
Online offer
Ends: 31-Dec-2026
30,485.00
Save 12,194.00 (40%)
Each
DNA Polymerase I/DNase I is an optimized mixture of both enzymes for efficient nick translation of DNA.

Application:
Labeling DNA with either radiolabeled or biotinylated nucleotides.

Source:
DNase I is purified from bovine pancreas; DNA Polymerase I from E. coli λ lysogen NM 964.

Performance and Quality Testing:
Performance tested in nick translation reaction.

Unit Definition:
One unit of DNA Polymerase I in the absence of DNase I incorporates 10 nmol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template•primer.

Unit Reaction Conditions:
50 mM potassium phosphate (pH 7.5), 6.7 mM MgCl2 , 1 mM 2-mercaptoethanol, 80 μg/ml template•primer, 32 μM dTTP, 69 nM [3H]dTTP, and enzyme in 100 μl for 30 min. at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionDNA Polymerase
Exonuclease Activity3' - 5', 5' - 3'
Hot StartNo
PolymeraseDNA Polymerase I
Quantity250 U
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
DNA Polymerase I/DNase I contains 0.5 units DNA Polymerase I and 0.4 milliunits DNase I per microliter. Store at -20°C.

Frequently asked questions (FAQs)

Does Thermo Fisher Scientific offer a protease-free DNase?

We do not test for protease activity as part of our QC but there is PMSF (a protease inhibitor) in the storage buffer. Furthermore, in the preparation of DNase I, we use a soybean trypsin inhibitor column to remove proteases.

Is there anything to prevent AmpliTaq Gold DNA polymerase from extending from the 3’ end of a TaqMan probe in a 5’ nuclease assay?

Yes. There is a phosphate group on the 3' end of all TaqMan probes that prevents such extension.

How does AmpliTaq Gold DNA Polymerase differ from AmpliTaq DNA Polymerase?

AmpliTaq Gold DNA Polymerase is a modified form of AmpliTaq DNA Polymerase that contains a proprietary chemical (or so-called hot start molecule) bound to the enzyme's active site. In order to activate the AmpliTaq Gold DNA Polymerase fully, we recommend an initial activation step of 95 degrees C for 10 min when using GeneAmp 10X PCR Buffer I and/or GeneAmp 10X PCR Buffer II and Mg in one of our thermal cyclers. When using GeneAmp 10X PCR Gold Buffer, activation time can be reduced to 5 minutes. Once activation is complete, you can proceed with your standard PCR cycling program (denaturing, annealing, extension, etc).

Does AmpliTaq Gold DNA Polymerase contain exonuclease (proofreading) activity?

No, AmpliTaq Gold DNA polymerase does not contain proofreading activity, however fidelity in PCR amplifications utilizing this enzyme may be improved. High fidelity can be achieved by: 1. Decreasing the final concentration of each nucleotide to 40-50 uM. 2. Using the lowest MgCl2 concentration possible. 3. Using less enzyme. 4. Decreasing extension times. 5. Using the highest annealing temperature possible. 6. Using as few cycles as possible.

Does the fidelity of AmpliTaq DNA Polymerase change in the presence of base analogs?

The fidelity of this PCR enzyme is affected in two ways. First, AmpliTaq DNA Polymerase typically binds to and incorporates base analogs less efficiently than conventional dNTPs, which means that polymerase activity is lower in reactions that contain base analogs. Second, the analog may pair with more than one conventional complementary template base, so the analog may be incorporated at an increased level compared to conventional dNTPs. For the best fidelity, we recommend that base analogs are included at low concentrations in the reaction.

Citations & References (1)

Citations & References
Abstract
Authors:
Journal:
PubMed ID:16195417