Orion™ Dissolved Oxygen Electrodes
Actual product may vary
Thermo Scientific™

Orion™ Dissolved Oxygen Electrodes

Use Thermo Scientific™ Orion™ Dissolved Oxygen Electrodes to assist in BOD measurements. Includes membrane module, O-ring and internal fill solution.
Have Questions?
Catalog NumberLength (English) Cable
181621X30 ft.
Catalog number 181621X
Price (INR)
-
Length (English) Cable:
30 ft.
Use Thermo Scientific™ Orion™ Dissolved Oxygen Electrodes to assist in BOD measurements. Includes membrane module, O-ring and internal fill solution.
Contains: Stainless steel housing, shielded cable and maintenance kit.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionDissolved Oxygen Electrode
For Use With (Equipment)Dissolved Oxygen Meters
Length (English) Cable30 ft.
Length (Metric) Cable9.14 m
TypeDissolved Oxygen Probe
Unit SizeEach