Pyridinium chlorochromate, 98%
Pyridinium chlorochromate, 98%
Pyridinium chlorochromate, 98%
Acros Organics

Pyridinium chlorochromate, 98%

CAS: 26299-14-9 | C5H6ClCrNO3 | 215.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 183670250
HSN 29333990
Price (INR)
4,876.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
4,876.00
Each
Chemical Identifiers
CAS26299-14-9
IUPAC Namehydrogen (trioxochromio)-λ³-chloranidyl pyridine
Molecular FormulaC5H6ClCrNO3
InChI KeyBUCMHZVAGRYTNK-UHFFFAOYSA-O
SMILES[H+].[Cl-][Cr](=O)(=O)=O.C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Iodometric97.5 to 102.5 %
Appearance (Form)Powder or crystals and/or chunks
Appearance (Color)Yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only