N-(tert-Butyl)hydroxylamine hydrochloride, 98%
N-(tert-Butyl)hydroxylamine hydrochloride, 98%
N-(tert-Butyl)hydroxylamine hydrochloride, 98%
Acros Organics

N-(tert-Butyl)hydroxylamine hydrochloride, 98%

CAS: 57497-39-9 | C4H11NO·HCl | 125.6 g/mol
Have Questions?
Catalog NumberQuantityPackaging
186910010
also known as 18691
1 gGlass bottle
Catalog number 186910010
also known as 18691
HSN 29280090
Price (INR)
7,954.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,954.00
Each
Chemical Identifiers
CAS57497-39-9
IUPAC Namehydrogen N-tert-butylhydroxylamine chloride
Molecular FormulaC4H12ClNO
InChI KeyDCSATTBHEMKGIP-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC(C)(C)NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point175°C to 185°C
Infrared spectrumConforms
Appearance (Form)Powder or crystals and/or chunks
Titration Argentometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only