Ethylenediamine dihydrochloride, 98+%
Ethylenediamine dihydrochloride, 98+%
Ethylenediamine dihydrochloride, 98+%
Acros Organics

Ethylenediamine dihydrochloride, 98+%

CAS: 333-18-6 | C2H10Cl2N2 | 133.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 187651000
HSN 29212100
Price (INR)
3,219.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
3,219.00
Each
Chemical Identifiers
CAS333-18-6
IUPAC Namedihydrogen ethane-1,2-diamine dichloride
Molecular FormulaC2H10Cl2N2
InChI KeyOHHBFEVZJLBKEH-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder, crystals or granules
Infrared spectrumConforms
Titration with NaOH>=98.0 %
pH3.9 to 5 (1 % in water)
Solubility(1 % in water) Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only