Levamisole hydrochloride, 99+%
Levamisole hydrochloride, 99+%
Levamisole hydrochloride, 99+%
Acros Organics

Levamisole hydrochloride, 99+%

CAS: 16595-80-5 | C11H12N2S·ClH | 240.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 187870100
also known as 18787
HSN 29349900
Price (INR)
9,074.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,074.00
Each
Chemical Identifiers
CAS16595-80-5
IUPAC Namehydrogen (6S)-6-phenyl-2H,3H,5H,6H-imidazo[2,1-b][1,3]thiazole chloride
Molecular FormulaC11H13ClN2S
InChI KeyLAZPBGZRMVRFKY-HNCPQSOCSA-N
SMILES[H+].[Cl-].C1CN2C[C@@H](N=C2S1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Loss on drying=<0.5 % (1 g, 105°C, 4 hrs)
Infrared spectrumConforms
Specific optical rotation-121° to -128° (20°C, 589 nm) (c=5, H2O)
Specific optical rotationon dried substance
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only