2-Iodothiophene, 98+%
2-Iodothiophene, 98+%
2-Iodothiophene, 98+%
Acros Organics

2-Iodothiophene, 98+%

CAS: 3437-95-4 | C4H3IS | 210.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
187880100
also known as 18788
10 mLGlass bottle
Catalog number 187880100
also known as 18788
HSN 29349900
Price (INR)
30,584.00
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
30,584.00
Each
Chemical Identifiers
CAS3437-95-4
IUPAC Name2-iodothiophene
Molecular FormulaC4H3IS
InChI KeyROIMNSWDOJCBFR-UHFFFAOYSA-N
SMILESIC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to red-brown to blue-green
Appearance (Form)Liquid
Refractive index1.6500 to 1.6540 (20°C, 589 nm)
GC>=98.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only