9-(Chloromethyl)anthracene, 98+%
9-(Chloromethyl)anthracene, 98+%
9-(Chloromethyl)anthracene, 98+%
Acros Organics

9-(Chloromethyl)anthracene, 98+%

CAS: 24463-19-2 | C15H11Cl | 226.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Packaging:
Glass bottle
Catalog number 188180100
also known as 18818
HSN 29039990
Price (INR)
31,909.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
31,909.00
Each
Chemical Identifiers
CAS24463-19-2
IUPAC Name9-(chloromethyl)anthracene
Molecular FormulaC15H11Cl
InChI KeyPCVRSXXPGXRVEZ-UHFFFAOYSA-N
SMILESClCC1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to golden
Infrared spectrumConforms
HPLC>=98.0 %
Melting point136°C to 141°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only