Sodium borohydride, 98+%, powder
Sodium borohydride, 98+%, powder
Sodium borohydride, 98+%, powder
Acros Organics

Sodium borohydride, 98+%, powder

CAS: 16940-66-2 | BH4Na | 37.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number 189300100
HSN 28500020
Price (INR)
6,036.00
Each
Quantity:
10 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (INR)
6,036.00
Each
Chemical Identifiers
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White
Titration Iodometric>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only