Tempshield™ Cryo Gloves - Elbow Length
Thermo Scientific™
Tempshield™ Cryo Gloves - Elbow Length
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
189565Large
189564Medium
Catalog number 189565
Size:
Large
Multi-layered construction allows for thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical. High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods. Safety certified to meet EU standards for CE Cat. III: EN511, EN388, EN ISO 21420. Available in wrist, elbow, mid-arm, shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
Specifications
DescriptionElbow Length
For Use With (Application)Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English)18 in.
Length (Metric)45.7 cm
ColorBlue
HandSpecific
SizeLarge
Unit SizeEach