Product Image
Thermo Scientific™
Forma™ Access Port for Refrigerators and Freezers
Provide a point of entry for probes and monitoring devices in lab refrigerators and freezes with the Thermo Scientific™ Forma™ Access Port for Refrigerators and Freezers.
Have Questions?
Catalog number 189620
Price (INR)
-
Provide a point of entry for probes and monitoring devices in lab refrigerators and freezes with the Thermo Scientific™ Forma™ Access Port for Refrigerators and Freezers.
Add an access port to a Forma refrigerator or freezer (chest or upright).
  • 1.3in. (0.7cm) inside diameter
  • Factory installed
  • Specify when ordering refrigerator or freezer
Specifications
DescriptionAccess Port
For Use With (Equipment)Refrigerators and Freezers
TypeFactory-installed
StyleUpright
Unit SizeEach