4-Methoxyphenylacetone, 98+%
4-Methoxyphenylacetone, 98+%
4-Methoxyphenylacetone, 98+%
Acros Organics

4-Methoxyphenylacetone, 98+%

CAS: 122-84-9 | C10H12O2 | 164.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 189800050
also known as 18980
HSN 29145000
Price (INR)
3,243.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
3,243.00
Each
Chemical Identifiers
CAS122-84-9
IUPAC Name1-(4-methoxyphenyl)propan-2-one
Molecular FormulaC10H12O2
InChI KeyWFWKNGZODAOLEO-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC(C)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5200 to 1.5300 (20°C, 589 nm)
Appearance (Color)Clear light yellow to yellow-green
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only