Gas Guard for CO<sub>2</sub> Tank
Thermo Scientific™
Gas Guard for CO2 Tank
For Use With: 3950/3951 Incubators
Have Questions?
Catalog NumberLighting
1900000LED
Catalog number 1900000
Price (INR)
-
Lighting:
LED
Minimize the need to frequently change out cylinders and reduce the risk of running out of gas. Thermo Scientific™ Gas Guards for CO2 and LN2 Tanks are factory installed and built in to specified CO2 incubators; specify when ordering the incubator. Fitting unobtrusively inside the incubator casing, these systems interface directly with unit power supply and control systems.
  • Factory-installed systems monitor CO2 or N2 pressure in source tanks
  • Automatically switch from one cylinder to another when supply in first tank is exhausted (pressure falls below 15psi)
  • Audible and visual alarms on incubator control panel alert to tank switch-overs and to when both tanks are low
Specifications
DescriptionGas Guard, CO2
LightingLED
TypeCO2 Gas Guard
Unit SizeEach