Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
Acros Organics

Germanium dioxide, 99.999%, (trace metal basis), electronic grade

CAS: 1310-53-8 | GeO2 | 104.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 190000500
HSN 28256010
Price (INR)
61,167.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
61,167.00
Each
Chemical Identifiers
CAS1310-53-8
IUPAC Namegermanedione
Molecular FormulaGeO2
InChI KeyYBMRDBCBODYGJE-UHFFFAOYSA-N
SMILESO=[Ge]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total trace metal impurities=<10 ppm
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only