Forma™ Filter Kit
Forma™ Filter Kit
Forma™ Filter Kit
Forma™ Filter Kit
Thermo Scientific™
Forma™ Filter Kit
For Use With: Forma Steri-cycle, Series 310 direct heat, Series II Water jacketed CO2 Incubator
Have Questions?
Catalog NumberType
1900067Filter Kit
Catalog number 1900067
Price (INR)
-
Type:
Filter Kit
Precisely control the CO2 and temperature with Thermo Scientific™ Forma™ Series CO2 Incubators. Forma™ CO2 Incubator Accessories are important components for optimal operation.
  • Precise control
  • Choice of TC or IR sensors
  • Superior parameter recovery characteristics
  • Continuous contamination control technology
Specifications
For Use With (Equipment)Forma™ Steri-Cycle™, Direct Heat and Water Jacket
LightingLED
TypeFilter Kit
CO2 Sensor TechnologyTC Sensor or IR Sensor
Unit SizeEach