Product Image
Thermo Scientific™
Solid Door Option for Small Chamber
Protection for your light sensitive samples
Have Questions?
Catalog NumberDoor Style
1900666TSSolid Door
Catalog number 1900666TS
Price (INR)
-
Door Style:
Solid Door
Solid outer door for Thermo Scientific™ chamber - replaces standard outer glass door to protect samples against light. Recommended for light sensitive applications. Factory-installed option, please order in conjunction with environmental chamber. F
  • Replaces standard outer glass door to protect samples against light
  • Recommended for light sensitive applications
  • Factory-installed option, please order in conjunction with environmental chamber
  • Choose based on size of chamber: small (for 311 L / 11 cu. ft. models) or large (for 821 L / 29 cu.ft. models)
  • Optional inner glass door enables visual check of samples with no major disturbance of temperature / humidity levels
Specifications
DescriptionSolid door for small chamber, replaces standard outer glass door, factory installed
Door StyleSolid Door
For Use With (Application)Light sensitive applications
For Use With (Equipment)Chamber models 3911, 3907
IncludesFactory installed solid door
LightingLED
TypeSolid Door Option
Unit SizeEach