2-Bromo-6-methoxynaphthalene, 98%
2-Bromo-6-methoxynaphthalene, 98%
2-Bromo-6-methoxynaphthalene, 98%
Acros Organics

2-Bromo-6-methoxynaphthalene, 98%

CAS: 5111-65-9 | C11H9BrO | 237.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 190560050
HSN 29093019
Price (INR)
4,924.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,924.00
Each
Chemical Identifiers
CAS5111-65-9
IUPAC Name2-bromo-6-methoxynaphthalene
Molecular FormulaC11H9BrO
InChI KeyAYFJBMBVXWNYLT-UHFFFAOYSA-N
SMILESCOC1=CC=C2C=C(Br)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
GC>=97.5 %
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point105°C to 111°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only