Aluminum tri-sec-butoxide, 97%
Aluminum tri-sec-butoxide, 97%
Aluminum tri-sec-butoxide, 97%
Acros Organics

Aluminum tri-sec-butoxide, 97%

CAS: 2269-22-9 | C12H27AlO3 | 246.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number 191215000
also known as 19121
HSN 29319090
Price (INR)
13,587.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
13,587.00
Each
Chemical Identifiers
CAS2269-22-9
IUPAC Namealuminium(3+) tris(butan-2-olate)
Molecular FormulaC12H27AlO3
InChI KeyWOZZOSDBXABUFO-UHFFFAOYNA-N
SMILES[Al+3].CCC(C)[O-].CCC(C)[O-].CCC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Titration Complexometric10.5 to 11.4 % (Al)
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only