Product Image
Gibco™

Fetal Bovine Serum, ultra-low IgG, NZ origin

Gibco fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays. It has earned the trust of researchers with consistent quality and award-winning support that helps meet research needs and budget requirements.
Have Questions?
Catalog NumberQuantity
1921005PJ500 mL
Catalog number 1921005PJ
Price (INR)
125,034.00
Each
Quantity:
500 mL

Gibco fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Specialty
  • Origin: New Zealand
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • IgG levels are less than 5 μg/mL and the BVD titer is low or not detectable.
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Shipping ConditionDry Ice
AgeFetal
Country of OriginNew Zealand
FormatBottle
Quantity500 mL
Serum TreatmentReduced IgG
Unit SizeEach
Contents & Storage
Store at -5°C to-20°C.

Frequently asked questions (FAQs)

What are the benefits and disadvantages of using heat-inactivated FBS in cell culture?

  1. Heating inactivates complement. Active complement can participate in cytolytic events, contract smooth muscle, release histamine from mast cells and platelets, and activate lymphocytic and macrophage cells. Applications where heat-inactivated serum is recommended include immunological studies and culturing of embryonic stem cells (ESCs), insect cells, and smooth muscle cells.
  2. Heat inactivation helps to achieve bottle-to-bottle and lot-to-lot stability by neutralizing many factors that can vary largely from lot to lot.
  3. There aren't necessarily disadvantages to heat inactivation of FBS, but there is some evidence that suggests there may be no added benefit to it unless you are carrying out immune studies.

Note: Heat inactivation is performed in a 56 degrees C water bath for 30 min with swirling every 10 min or so for heat distribution and to lower the degree of protein aggregation/flocculant precipitation. Note: If the time or temperature is exceeded, the serum may thicken to a gel. If this occurs, the serum is no longer usable. Unnecessary heat inactivation can take up time and potentially lead to wasted reagents if a mistake is made during the protocol1.

1. Pellerin, et al., Bioengineering, published in 2021.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.