Lithium fluoride, 99.98%, extra pure
Lithium fluoride, 99.98%, extra pure
Lithium fluoride, 99.98%, extra pure
Acros Organics

Lithium fluoride, 99.98%, extra pure

CAS: 7789-24-4 | FLi | 25.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Plastic bottle
Catalog number 193380250
HSN 28261990
Price (INR)
28,098.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
28,098.00
Each
Chemical Identifiers
CAS7789-24-4
IUPAC Namelithium(1+) fluoride
Molecular FormulaFLi
InChI KeyPQXKHYXIUOZZFA-UHFFFAOYSA-M
SMILES[Li+].[F-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total trace metal impurities=<0.02 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only