Mercury(II) oxide, 99+%, ACS reagent, red
Mercury(II) oxide, 99+%, ACS reagent, red
Mercury(II) oxide, 99+%, ACS reagent, red
Acros Organics

Mercury(II) oxide, 99+%, ACS reagent, red

CAS: 21908-53-2 | HgO | 216.59 g/mol
Have Questions?
Catalog NumberQuantityPackaging
193511000100 gGlass bottle
Catalog number 193511000
HSN 98020000
Price (INR)
16,097.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
16,097.00
Each
Chemical Identifiers
CAS21908-53-2
IUPAC Nameoxomercury
Molecular FormulaHgO
InChI KeyUKWHYYKOEPRTIC-UHFFFAOYSA-N
SMILESO=[Hg]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=99.0 %
Insoluble matter=<0.03 % (in dilute HCl)
Residue after reduction=<0.025 %
Nitrogen compounds (as N)=<0.005 %
Appearance (Color)Orange-red to red
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only