Nickel, 99.9%, (trace metal basis), powder, -325 mesh, Thermo Scientific Chemicals
Nickel, 99.9%, (trace metal basis), powder, -325 mesh, Thermo Scientific Chemicals
Nickel, 99.9%, (trace metal basis), powder, -325 mesh, Thermo Scientific Chemicals
Acros Organics

Nickel, 99.9%, (trace metal basis), powder, -325 mesh, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
193610250
also known as 19361
25 gPlastic bottle
Catalog number 193610250
also known as 19361
HSN 28051900
Price (INR)
6,510.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
6,510.00
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total trace metal impurities=<0.1 %
Particle size=<44 µm
Appearance (Color)Grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only