Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)
Acros Organics

Ammonium dihydrogen phosphate, 99.9%, (trace metal basis)

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
193700025
also known as 19370
2.5 kgPlastic bottle
Catalog number 193700025
also known as 19370
HSN 28352990
Price (INR)
30,039.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
30,039.00
Each
Chemical Identifiers
CAS7722-76-1
IUPAC Namephosphoric acid amine
Molecular FormulaH6NO4P
InChI KeyLFVGISIMTYGQHF-UHFFFAOYSA-N
SMILESN.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<0.1 %
Appearance (Color)White
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only