Replacement Screws for MaxQ™ Shakers
Thermo Scientific™
Replacement Screws for MaxQ™ Shakers
Use these replacement screws for Thermo Scientific™ MaxQ™ shakers with the clamps that secure vessels to the universal shaker platform.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
194046Clamps for MaxQ Shaker Platform
194024Tube Holders for MaxQ Shaker Platform
Catalog number 194046
Price (INR)
-
For Use With (Equipment):
Clamps for MaxQ Shaker Platform
Use Thermo Scientific™ Replacement Screws for MaxQ™ Shakers with the clamps that secure vessels to the universal shaker platform.
Specifications
DescriptionPhillips head screws
For Use With (Equipment)Clamps for MaxQ Shaker Platform
Packaging Quantity50/Pk.
Product TypePhillips Head Screws
Unit SizeEach