Product Image
Gibco™
Donor Horse Serum, heat inactivated, New Zealand origin (custom product)
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers withRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1941300PP4 L
1941300PG100 mL
Catalog number 1941300PP
Price (INR)
-
Quantity:
4 L
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
Sera Category: Specialty
This is a GIBCO Custom Product. Please complete and submit the Custom Media Inquiry Form and a Customer Service Representative will contact you regarding pricing and availability.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
SpeciesHorse/Equine, Horse
Country of OriginNew Zealand
Quantity4 L
Serum TreatmentHeat-inactivated
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C)
Frequently asked questions (FAQs)
When should I use heat-inactivated serum?
Heat-inactivation of serum inactivates the “complements” in the serum. In general, it is not necessary to use heat-inactivated serum to culture most cell types. We recommend using heat-inactivated serum when working with immune type cells or for immunological applications.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.