CaptureSelect™ FSH Affinity Matrix
Thermo Scientific™

CaptureSelect™ FSH Affinity Matrix

Thermo Scientific™ CaptureSelect™ FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1943180250250 mL
1943180055 mL
19431801010 mL
19431805050 mL
19431801L1 L
19431805L5 L
Catalog number 1943180250
Price (INR)
-
Quantity:
250 mL

CaptureSelect FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications
Flow Rate200 to 400 cm/hr.
MatrixAgarose-based, aldehyde-activated
Quantity250 mL
Stationary PhaseAnti-Follicle Stimulating Hormone
FormLiquid Suspension
Particle Size65 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).