CaptureSelect™ hCG Affinity Matrix
Thermo Scientific™

CaptureSelect™ hCG Affinity Matrix

Thermo Scientific™ CaptureSelect™ hCG Affinity Matrix was specifically designed for the purification of human chorionic gonadotropin (hCG) from recombinant sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
19434105L5 L
1943410055 mL
19434101010 mL
194341050
also known as 1943410050
50 mL
1943410250250 mL
19434101L1 L
Catalog number 19434105L
Price (INR)
-
Quantity:
5 L

CaptureSelect hCG Affinity Matrix was specifically designed for the purification of human chorionic gonadotropin (hCG) from recombinant sources.

Some features of the CaptureSelect hCG Affinity Matrix include:

  • Selective for hCG, with cross-binding to LH, FSH, and TSH
  • Enables efficient purification of recombinant gonadotropins with high yields and purity
  • Offers excellent scalability
  • Non–animal-derived

Human gonadotropins (hCG, LH, FSH, TSH) are glycoprotein hormones secreted by gonadotrope cells of the pituitary gland that are often produced as recombinant proteins. They have an identical alpha chain subunit that is recognized by the ligand. Consequently, the CaptureSelect hCG Affinity Matrix shows binding to all human gonadotropins. The high selectivity and yields obtained using CaptureSelect hCG Affinity Matrix enable a robust and efficient purification process for hCG with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect hCG affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: approx. 4 mg/mL resin (determined with HCG)
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
Specifications
Binding Capacity4 mg/mL
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeHCG affinity ligand
MatrixAgarose-based, aldehyde-activated
Particle Size65 ± 10 μm
Quantity5 L
Shipping ConditionRoom Temperature
Stationary PhaseAnti-gonadotropin
FormAgarose Bead
Particle Size65 ±10 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).