Product Image
Gibco™

Fibrinogen Bovine Protein, Freeze Dried

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1949400IM100 g
1949400II1 g
Catalog number 1949400IM
Price (INR)
-
Quantity:
100 g
This is a GIBCO Custom Product. Please complete and submit the Custom Media Inquiry Form and a Customer Service Representative will contact you regarding pricing and availability.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gene AliasBovine Fibrinogen
Product LineGibco
Protein SubtypeFibrinogen
Quantity100 g
Research CategoryExtracellular Matrix & Cell Adhesion
Shipping ConditionRoom Temperature
For Use With (Application)Cell Culture
SpeciesCattle/Bovine
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).