Product Image
Thermo Scientific™
Hot Gas Bypass Temperature Controllers
Combine these hot gas bypass temperature controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVoltage
195023Factory-Installed220 V
195021Factory-Installed120 V
195700Field-Installed120 V
195701Field-Installed220 V
Catalog number 195023
Price (INR)
-
Type:
Factory-Installed
Voltage:
220 V
Combine Thermo Scientific™ Hot Gas Bypass Temperature Controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
Features:
  • 120V or 220V
  • Factory or field installed
Ordering Information:
Field installed option must be installed by a qualified professional.
  • Warranty varies by country, please contact us for details
  • Specifications
    DescriptionFactory Installed
    For Use With (Equipment)CryoPlus 1,2,3,4
    TypeFactory-Installed
    Voltage220 V
    Unit SizeEach