Product Image
Thermo Scientific™
Sliding Drawer Inserts
Maximize storage space with these sliding drawer inserts, for use with racks used in Thermo Scientific™ 13, 17, 23 and 28 cu. ft. ultra-low temperature freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
1950666P513 cu. ft. FreezersMicroplate Insert
1950668P513 cu. ft. Freezers2 in. Box Insert
1950669P517 and 23 cu. ft. FreezersMicroplate Insert
1950670P517 and 23 cu. ft. Freezers2 in. Box Insert
1950671P528 cu. ft. FreezersMicroplate Insert
1950672P528 cu. ft. Freezers2 in. Box Insert
Catalog number 1950666P5
Price (INR)
-
For Use With (Equipment):
13 cu. ft. Freezers
Type:
Microplate Insert
Maximize storage space with Thermo Scientific™ Sliding Drawer Inserts, for use with racks used in Thermo Scientific™ 13, 17, 23 and 28 cu. ft. ultra-low temperature freezers.
  • Warranty varies by country, please contact us for details
Specifications
For Use With (Equipment)13 cu. ft. Freezers
HoldsMicroplate Rack
TypeMicroplate Insert
Capacity (English)28 cu. ft.
Capacity (Metric)792.8 L
Unit SizeEach