Biotin-14-CTP is a CTP analog with biotin attached at the N 4 -position of the pyrimidine base by a 14-atomRead more
Have Questions?
Catalog Number
Quantity
19519016
500 nmol
Catalog number 19519016
Price (INR)
54,946.00
Offre exceptionnelle en ligne
Ends: 31-Mar-2026
91,576.00
Save 36,630.00 (40%)
Each
Quantity:
500 nmol
Price (INR)
54,946.00
Offre exceptionnelle en ligne
Ends: 31-Mar-2026
91,576.00
Save 36,630.00 (40%)
Each
Biotin-14-CTP is a CTP analog with biotin attached at the N 4 -position of the pyrimidine base by a 14-atom linker. Biotin-14-CTP can be efficiently incorporated into RNA transcripts using T7, SP6, or T3 RNA polymerase and the appropriate template in the presence of ATP, GTP, UTP, and Biotin-14-CTP (1). The biotinylated RNA can be used as a probe in membrane-based or in situ hybridizations (2). The amount of material provided is sufficient for up to 20 transcription reactions (1 μg template).
Performance and Quality Testing: Purity is evaluated by reverse-phase HPLC.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration10 mM
FormLiquid
Label or DyeBiotin & Other Hapten
Product TypeLabeled Nucleotide
Quantity500 nmol
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Biotin-14-CTP is supplied at 10 mM in 10 mM Tris-HCl (pH 7.5), 0.1 mM EDTA. Store at -20°C.