Palladium(II) chloride, 59% Pd
Palladium(II) chloride, 59% Pd
Palladium(II) chloride, 59% Pd
Acros Organics

Palladium(II) chloride, 59% Pd

CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 195200010
HSN 28439019
Price (INR)
31,625.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
31,625.00
Each
Chemical Identifiers
CAS7647-10-1
IUPAC Namepalladium(2+) dichloride
Molecular FormulaCl2Pd
InChI KeyPIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Copper (Cu)=<15 ppm
Magnesium (Mg)=<15 ppm
Silicon (Si)=<30 ppm
Appearance (Color)Red-brown
Calcium (Ca)=<30 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only