Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
Acros Organics

Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH

CAS: 7440-38-2 | As | 74.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number 195825000
also known as 19582
HSN 28048000
Price (INR)
18,014.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (INR)
18,014.00
Each
Chemical Identifiers
CAS7440-38-2
IUPAC Namearsenic
Molecular FormulaAs
InChI KeyRQNWIZPPADIBDY-UHFFFAOYSA-N
SMILES[As]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Strength997 to 1003 ug/ml of As(3+) in 2% KOH (20°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only