Cadmium standard solution, for AAS, 1 mg/ml Cd in 2 to 5% HNO3
Cadmium standard solution, for AAS, 1 mg/ml Cd in 2 to 5% HNO3
Cadmium standard solution, for AAS, 1 mg/ml Cd in 2 to 5% HNO3
Acros Organics

Cadmium standard solution, for AAS, 1 mg/ml Cd in 2 to 5% HNO3

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number 195875000
HSN 28051900
Price (INR)
14,298.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (INR)
14,298.00
Each
Chemical Identifiers
CAS10022-68-1
IUPAC Namecadmium(2+) bis(nitric acid)
Molecular FormulaCdH2N2O6
InChI KeyIZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Strength990 to 1010 µg/ml of Cd(2+) in 2 to 5% HNO3
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only