Product Image
Thermo Scientific™
Forma™ Sample Probe Kits
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
195892Sample Probe, Customer Installed
195893Sample Probe, Factory Installed
Catalog number 195892
Price (INR)
-
Type:
Sample Probe, Customer Installed
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits. Use probe kit with Forma 8600 Series Upright Freezers.
  • Useful for measuring sample temperature or top chamber temperature
  • Kits consist of probe and probe kit
Specifications
TypeSample Probe, Customer Installed
StyleUpright
Unit SizeEach